missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Nalgene™ Unitary™ LDPE Wash Bottles

Description
Dispense contents without tipping or shaking with Thermo Scientific™ Nalgene™ Unitary™ LDPE Wash Bottles. Wide mouth allows faster, easier filling. Bottle and tubulation are molded in one piece for uniform service and precision flow without sputtering. The tip can be cut back to increase flow. Made from high-quality, laboratory-grade, plastic materials for dependably low leachables and extractables. Naturally flexible LDPE material is chemically compatible with a wide range of laboratory reagents and provides an ergonomic soft squeeze to help reduce hand fatigue.
Features:
- Bottle and tubulation are molded in one piece for uniform leakproof† service
- Tip delivers a uniform, ultrafine stream without sputtering, but can also be cut back to increase flow
- Leakproof†
Includes:
Polypropylene screw closure
Specifications
Specifications
| Capacity | 4 oz., 125 mL |
| Capacity (Metric) | 125 mL |
| Capacity (English) | 4 oz. |
| Color | Translucent |
| Description | LDPE bottle/tubulation |
| Material | LDPE |
| No. per Case | 48 |
| No. per Pack | 6 |
| Closure Included | Yes |
| Graduated | No |
| Show More |