missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Unitary™ LDPE Wash Bottles
GREENER_CHOICE

Catalog No. 0340910EE
Change view
Click to view available options
Capacity (Metric):
125 mL
250 mL
500 mL
750 mL
1000 mL
Closure Size:
24 mm
38 mm
43 mm
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Closure Size
0340910EE 750 mL 38 mm
0340910C 125 mL 24 mm
0340910D 250 mL 24 mm
0340910E 500 mL 38 mm
0340910F 1000 mL 43 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 0340910EE Supplier Thermo Scientific™ Supplier No. 24020750
Only null left

Dispense contents without tipping or shaking by using these LDPE wash bottles, designed with a wide mouth for faster, easier filling.

Dispense contents without tipping or shaking with Thermo Scientific™ Nalgene™ Unitary™ LDPE Wash Bottles. Wide mouth allows faster, easier filling. Bottle and tubulation are molded in one piece for uniform service and precision flow without sputtering. The tip can be cut back to increase flow. Made from high-quality, laboratory-grade, plastic materials for dependably low leachables and extractables. Naturally flexible LDPE material is chemically compatible with a wide range of laboratory reagents and provides an ergonomic soft squeeze to help reduce hand fatigue.

Features:
  • Bottle and tubulation are molded in one piece for uniform leakproof† service
  • Tip delivers a uniform, ultrafine stream without sputtering, but can also be cut back to increase flow
  • Leakproof†

Includes:

Polypropylene screw closure

Specifications

Capacity 24 oz., 750 mL
Capacity (Metric) 750 mL
Capacity (English) 24 oz.
Color Translucent
Description LDPE bottle/tubulation
Material LDPE
No. per Case 24
No. per Pack 4
Closure Included Yes
Graduated No
Mouth Wide
Shape Cylindrical
Height (Metric) 204.5 mm
Product Type Wash Bottle
Height (English) 8.05 in.
Closure Material Polypropylene
Closure Size 38 mm
Diameter (Metric) 84.8 mm
Closure Type Screw Cap
Through the Closure Dispensing No
Show More Show Less