Learn More
Thermo Scientific™ Nalgene™ Square Wide-Mouth PPCO Bottles with Closure: Bulk Pack, Autoclavable
GREENER_CHOICE
Save valuable storage space with these easy-to-grip, square, wide-mouth PPCO bottles, which feature durable, uniform walls resistant to splitting or puncturing.
$3,796.63
Specifications
Color Translucent
Description Wide-Mouth Square Bottle
Material PPCO
Sterility Non-sterile
Closure Color Natural
View More Specs
Products 1
Catalog Number Mfr. No. Capacity (Metric) Closure Size Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Closure Size Price Quantity & Availability  
02923803
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
3121100008
250 mL 43 mm
Case of 250 Each for $3,796.63
Only null left
Description
Description
Save valuable storage space with the easy-to-grip Thermo Scientific™ Nalgene™ Square Wide-Mouth PPCO Bottles with Closure. These bottles feature durable, uniform walls resistant to splitting or puncturing.
Features:
  • Polypropylene copolymer
  • Wide mouth for easy filling and pouring
  • Leakproof†, linerless polypropylene caps
  • Autoclavable
Includes:
Linerless polypropylene closures that ensure leakproof† performance without a liner that can wrinkle, cause leaks and contaminate solutions
Ordering Information:
Bulk packed in case; closures included but not assembled.
Specifications
Specifications
Translucent
PPCO
Natural
Wide
Unlined
Screw Cap
Autoclavable
Wide-Mouth Square Bottle
Non-sterile
Yes
Square
Polypropylene
No
Videos
SDS
Documents
Documents
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.