missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Square Wide-Mouth HDPE Bottles with Closure
GREENER_CHOICE

Catalog No. 03312B
Change view
Click to view available options
Capacity (Metric):
60 mL
175 mL
250 mL
500 mL
1 L
Closure Size:
28 mm
38 mm
43 mm
53 mm
63 mm
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Closure Size
03312B 250 mL 43 mm
03312AA 60 mL 28 mm
03312A 175 mL 38 mm
03312C 500 mL 53 mm
03312D 1 L 63 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 03312B Supplier Thermo Scientific™ Supplier No. 21140008
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Easily fill these square, wide-mouth HDPE bottles that offer excellent chemical resistance, making them suitable for a wide variety of uses.

Easily fill Thermo Scientific™ Nalgene™ Square Wide-Mouth HDPE Bottles. HDPE exhibits excellent chemical resistance to most acids, bases and alcohols, making these bottles suitable for a wide variety of uses. Good for freezer storage to -100°C (-148°F). This product is ACT Labeled by My Green Lab to enable data-based sustainable product purchasing decisions.

  • Excellent chemical resistance to most acids, bases and alcohols make these excellent containers to protect your valuable samples
  • Rectangular shape makes it easy to fit in freezer for storage up to -100°C (-148°F)
  • Linerless polypropylene screw closures provide the ultimate in leakproof† protection without the use of a liner that can wrinkle, cause leaks, or contaminate your reagents
  • Leakproof†

Includes:

Polypropylene screw caps

Recycling Information: Nalgene reusable HDPE bottles are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually. Nalgene HDPE bottles and their packaging are readily recyclable. Please recycle the corrugate cartons and LDPE bags in which your Nalgene bottles products are packaged. And at end of product life, please wash and recycle your Nalgene HDPE bottles.

Specifications

Capacity 8 oz., 250 mL
Capacity (Metric) 250 mL
Color Translucent
Description Wide-Mouth Square Bottle
Diameter (English) 2.4 in.
Diameter (English) Inner Neck 1.3 in.
Diameter (Metric) Inner Neck 33 mm
Material HDPE
No. per Case 72
No. per Pack 12
Sterility Non-sterile
Closure Color Natural, translucent
Closure Included Yes
Graduated No
Mouth Wide
ACT Label Yes
Height (Metric) 116 mm
Width (English) 2.4 in.
Height (English) 4.5 in.
Closure Liner Unlined
Closure Material Polypropylene
Closure Size 43 mm
Diameter (Metric) 61 mm
Closure Type Screw Cap
Through the Closure Dispensing No
Capacity (English) 8 oz.
Shape Square
Certifications/Compliance USP Class VI; FDA - 21 CFR
Product Type Square Wide Mouth Bottle
Show More Show Less

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.