missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Square PETG Media Bottles with Septum Closure: Sterile, Shrink-Wrapped Trays
GREENER_CHOICE

Catalog No. 03313900
Change view
Click to view available options
Capacity (Metric):
500 Expanded
60 mL
125 mL
500 mL
1 L
Closure Size:
24 mm
38-430 mm
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Closure Size
03313900 60 mL 24 mm
03313901 125 mL 38-430 mm
03313902 500 mL 38-430 mm
03313903 1 L 38-430 mm
14178159 500 Expanded 38-430 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 03313900 Supplier Thermo Scientific™ Supplier No. 3420230060

Package biological and diagnostic reagents that require aseptic dispensing or addition of supplements by syringe in these durable, break-resistant media bottles.

Thermo Scientific™ Nalgene Square PETG Media Bottles are durable, break-resistant, and feature a septum closure for aseptic syringe dispensing or addition of supplements.

Features:
  • HDPE closure with silicone/PTFE septum provides a PTFE fluid contact surface that is inert and nonstick to ensure fluid integrity.

    • Leakproof†, transparent media bottles
    • Square design saves valuable shelf space
    • Bottles and closures are sterile to 10-6 sterility assurance level

    Ordering Information:

    Packed in shrink-wrapped trays in double-polybagged cartons for cleanroom applications.

    Recommended for:

    Ideal for packaging of biological and diagnostic reagents that require aseptic dispensing or addition of supplements by syringe.

    Note:

    † The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Specifications

Capacity (Metric) 60 mL
Capacity (English) 2 oz.
Color Transparent
Description With septum closure
Diameter (English) 1.6 in.
Diameter (English) Inner Neck 0.71 in.
Diameter (Metric) Inner Neck 18 mm
Material PETG
No. per Case 200
No. per Pack 40
Sterility Sterile
Certifications/Compliance Meet USP Class IV standards. Noncytotoxic.
Closure Included Yes
Graduated Yes
Mouth Wide
Shape Square
Fit for Purpose Sterile
Height (Metric) 82 mm
Product Type Media Bottle
Height (English) 3.23 in.
Closure Size 24 mm
Diameter (Metric) 41 mm
Show More Show Less