missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Square PET Media Bottles with Closure: Sterile, Shrink-Wrapped Trays
GREENER_CHOICE

Catalog No. 0339072
Change view
Click to view available options
Capacity (Metric):
125 mL
250 mL
650 mL
1 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
0339072 125 mL
02923145 250 mL
0292500A 650 mL
0292500B 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 0339072 Supplier Thermo Scientific™ Supplier No. 3420400125
Edge

missing translation for 'validMainframeMsg'

Provide excellent gas barrier properties for storage and shipping of liquid media, buffers and sera with these square bottles.

Provide excellent gas barrier properties for storage and shipping of liquid media, buffers and sera with Thermo Scientific™ Nalgene™ Square PET Media Bottles with Closure. These light-weight bottles are shatter-resistant, sterile and come in a space-saving square design.

Features:
  • PET with natural HDPE closure
  • Space saving square design
  • Molded in graduations and neck ring for use with tamper evident shrink sleeves
  • Sterile, non-pyrogenic, non-cytotoxic
  • Excellent gas barrier, with transparency and reduced permeability to CO2 and O2
  • Bottle and closure material are ADCF (Animal Derived Component Free)
  • Leakproof†

Recommended for: Storage and shipping of liquid media, buffers and sera.

Ordering Information:

Products are packaged in shrink-wrap trays. Trays are double polybagged inside the master carton. † The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes; b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Specifications

Capacity (Metric) 125 mL
Color Clear
Diameter (English) 2.04 in.
Diameter (English) Inner Neck 1.1 in.
Diameter (Metric) Inner Neck 28 mm
Material PET
No. per Case 96
No. per Pack 24
Sterility Sterile
Closure Included Yes
Graduated Yes
Mouth Narrow
Fit for Purpose Sterile
Height (Metric) 104 mm
Height (English) 4.09 in.
Closure Size 38-430 mm
Diameter (Metric) 52 mm
Capacity (English) 4.2 oz.
Shape Square
Certifications/Compliance PET Media Bottles and HDPE closure materials meet current biological testing standards including USP Class VI, USP <661.1>
Product Type Media Bottle
Show More Show Less

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for two minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.