Learn More
Thermo Scientific™ Nalgene™ Square Narrow-Mouth HDPE Bottles with Closure

Conserve space with these square, narrow-mouth HDPE bottles that are chemically and break resistant, as well as economically priced for a wide range of lab uses.
Supplier: Thermo Scientific™ 20180250
Description
Conserve space with Thermo Scientific™ Nalgene™ Square Narrow-Mouth HDPE Bottles with Closure. HDPE bottles are the most chemically and break resistant of all Nalgene square bottles, yet are economically priced for a wide range of lab uses. They are made only from high-quality, laboratory-grade plastic materials for dependably low leachables and extractables. This product is ACT Labeled by My Green Lab to enable data-based sustainable product purchasing decisions.
Features:
- Square sides allow for good use of space and an easy grip
- Caps provide leakproof† protection without the use of a liner that can wrinkle, cause leaks or contaminate your reagents
- Graduated for easy dispensing
- Leakproof† when used with Nalgene closures
- Can be used to -100°C (-148°F)
- Not autoclavable
- Participates in My Green Lab's ACT Label program for Accountability, Consistency and Transparency (ACT) around evaluating product sustainability
- Bottle and carton are readily recyclable in most communities
- Manufactured from non-toxic materials
Includes:
Leakproof† Polypropylene Screw Caps
Note:
Nalgene LDPE, HDPE, PP and PMP containers can be used for long-term storage, but direct UV exposure should be avoided.
Recycling Information:
Nalgene reusable HDPE bottles are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually. Nalgene HDPE bottles and their packaging are readily recyclable. Please recycle the corrugate cartons and LDPE bags in which your Nalgene bottles products are packaged. And at end of product life, please wash and recycle your Nalgene HDPE bottles.
Chemical Identifiers
| Translucent | |
| Leakproof polypropylene screw closures | |
| Non-sterile | |
| Yes | |
| Yes | |
| Narrow | |
| Yes | |
| Polypropylene | |
| No |
| Narrow-Mouth Square Bottle | |
| HDPE | |
| Natural, translucent | |
| USP Class VI, FDA - 21 CFR | |
| Yes | |
| Square | |
| Square Narrow Mouth Bottle | |
| Screw Cap | |
| Not Autoclavable |
Specifications
| 8 oz., 250 mL | |
| 8 oz. | |
| Narrow-Mouth Square Bottle | |
| 1.1 in. | |
| Leakproof polypropylene screw closures | |
| 72 | |
| Non-sterile | |
| Yes | |
| Yes | |
| Narrow | |
| Yes | |
| Square Narrow Mouth Bottle | |
| 5.7 in. | |
| 38-430 | |
| Screw Cap | |
| Not Autoclavable |
| 250 mL | |
| Translucent | |
| 2.4 in. | |
| 28 mm | |
| HDPE | |
| 12 | |
| Natural, translucent | |
| USP Class VI, FDA - 21 CFR | |
| Yes | |
| Square | |
| 146 mm | |
| 2.41 in. | |
| Polypropylene | |
| 61 mm | |
| No |
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.