missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Square Narrow-Mouth HDPE Bottles with Closure
GREENER_CHOICE

Catalog No. 033111N
Change view
Click to view available options
Capacity (Metric):
30 mL
60 mL
125 mL
250 mL
1 L
Closure Size:
38-430
20 mm
24 mm
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Closure Size
033111N 30 mL 20 mm
033111P 60 mL 24 mm
033111Q 125 mL 38-430
033111R 250 mL 38-430
033111T 1 L 38-430
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 033111N Supplier Thermo Scientific™ Supplier No. 20180030

Conserve space with these square, narrow-mouth HDPE bottles that are chemically and break resistant, as well as economically priced for a wide range of lab uses.

Conserve space with Thermo Scientific™ Nalgene™ Square Narrow-Mouth HDPE Bottles with Closure. HDPE bottles are the most chemically and break resistant of all Nalgene square bottles, yet are economically priced for a wide range of lab uses. They are made only from high-quality, laboratory-grade plastic materials for dependably low leachables and extractables. This product is ACT Labeled by My Green Lab to enable data-based sustainable product purchasing decisions.

Features:

  • Square sides allow for good use of space and an easy grip
  • Caps provide leakproof† protection without the use of a liner that can wrinkle, cause leaks or contaminate your reagents
  • Graduated for easy dispensing
  • Leakproof† when used with Nalgene closures
  • Can be used to -100°C (-148°F)
  • Not autoclavable
  • Participates in My Green Lab's ACT Label program for Accountability, Consistency and Transparency (ACT) around evaluating product sustainability
  • Bottle and carton are readily recyclable in most communities
  • Manufactured from non-toxic materials

Includes:

Leakproof† Polypropylene Screw Caps

Note:

Nalgene LDPE, HDPE, PP and PMP containers can be used for long-term storage, but direct UV exposure should be avoided.

Recycling Information:

Nalgene reusable HDPE bottles are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually. Nalgene HDPE bottles and their packaging are readily recyclable. Please recycle the corrugate cartons and LDPE bags in which your Nalgene bottles products are packaged. And at end of product life, please wash and recycle your Nalgene HDPE bottles.

Specifications

Capacity 1 oz., 30 mL
Capacity (Metric) 30 mL
Capacity (English) 1 oz.
Color Translucent
Description Narrow-Mouth Square Bottle
Diameter (English) 1.5 in.
Diameter (English) Inner Neck 0.5 in.
Diameter (Metric) Inner Neck 14 mm
Includes Leakproof polypropylene screw closures
Material HDPE
No. per Case 72
No. per Pack 12
Sterility Non-sterile
Closure Color Natural, translucent
Graduations Yes
Certifications/Compliance USP Class VI, FDA - 21 CFR
Closure Included Yes
Graduated Yes
Mouth Narrow
Shape Square
ACT Label Yes
Height (Metric) 64 mm
Product Type Square Narrow Mouth Bottle
Width (English) 1.52 in.
Height (English) 2.5 in.
Closure Material Polypropylene
Closure Size 20 mm
Diameter (Metric) 38 mm
Closure Type Screw Cap
Through the Closure Dispensing No
Autoclavable Not Autoclavable
Show More Show Less

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.