Learn More
Thermo Scientific™ Nalgene™ Spigots for Storage Tanks
Ensure safe and efficient liquid dispensing with Thermo Scientific™ Nalgene™ Spigots for Storage Tanks, for use with Naglene™ tanks up to 100 gal. with factory installed threaded boss.
Supplier:  Thermo Scientific™ 64200750
 View more versions of this product
Catalog No. 02-963-7B
Only null left
Description
Description
Ensure safe and efficient liquid dispensing from tanks by using Thermo Scientific™ Nalgene™ Spigots for Storage Tanks. Spigots are for use with Naglene™ tanks up to 100 gal. with factory installed threaded boss.
Includes
  • PTFE O-rings (6421-0010 includes two O-rings)
Note:
Make sure container and spigot have compatible chemical resistance before installing.
Specifications
Specifications
Spigot
LDPE
378 L
1
19mm NPT male screw thread
100 gal.
6
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
Disclaimer to internal research use restriction:
Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.