Learn More
Thermo Scientific™ Nalgene™ Sealing Caps for Oak Ridge Centrifuge Tubes; PP screw closure, silicone gasket
Ensure leakproof service at their maximum speeds >10,000 x g, even when full
$248.63
| Catalog Number | Mfr. No. | For Use With (Equipment) | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | For Use With (Equipment) | Price | Quantity & Availability | |||||
02945124
![]() |
Thermo Scientific™
7131320058 |
Replacement O-ring (for Mfr. No. DS3132-0058) |
Case of 10 Each for $248.63
|
|
|||||
Description
- Polypropylene closures with silicone gasket
Before autoclaving, set closure on top of the container without engaging the threads.
Specifications
| Autoclavable | |
| Silicone | |
| Silicone |