missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Rectangular HDPE Bottles with Closure
GREENER_CHOICE

Catalog No. 029245G
Change view
Click to view available options
Capacity (Metric):
125 mL
250 mL
500 mL
1 L
2 L
Closure Size:
28 mm
38 mm
48 mm
53 mm
63 mm
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Closure Size
029245G 1 L 53 mm
029245A 125 mL 28 mm
029245C 250 mL 38 mm
029245E 500 mL 48 mm
029245J 2 L 63 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 029245G Supplier Thermo Scientific™ Supplier No. 20070032
Edge

Transport, sample or store liquids and dry materials with these space-efficient bottles featuring excellent chemical resistance to most acids, bases and alcohols.

Transport, sample or store liquids and dry materials with these space-efficient bottles that feature excellent chemical resistance to most acids, bases and alcohols. Thermo Scientific™ Nalgene™ Rectangular HDPE Bottles with Closure take up less shelf space than traditional round bottles. HDPE bottles have a sturdy wall construction with straight sides and a wide mouth for easy filling. Made only from high-quality, laboratory-grade, plastic materials for dependably low leachables and extractables. This product is ACT Labeled by My Green Lab to enable data-based sustainable product purchasing decisions.

Features:

  • Excellent chemical resistance to most acids, bases and alcohols make these excellent containers to protect your valuable samples
  • Rectangular shape makes it easy to fit in freezer for storage up to -100°C (-148°F)
  • Linerless polypropylene screw closures provide the ultimate in leakproof† protection without the use of a liner that can wrinkle, cause leaks, or contaminate your reagents
  • Leakproof†

Includes:

Polypropylene screw caps

Recycling Information: Nalgene reusable HDPE bottles are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually. Nalgene HDPE bottles and their packaging are readily recyclable. Please recycle the corrugate cartons and LDPE bags in which your Nalgene bottles products are packaged. And at end of product life, please wash and recycle your Nalgene HDPE bottles.

Specifications

Capacity 32 oz., 1000 mL
Capacity (Metric) 1 L
Color Translucent
Description Wide-Mouth Rectangular Bottle
Diameter (English) Inner Neck 1.7 in.
Diameter (Metric) Inner Neck 44 mm
Material HDPE
Length (Metric) 125 mm
No. per Case 24
No. per Pack 6
Sterility Non-sterile
Width (Metric) 71 mm
Closure Color Natural, translucent
Closure Included Yes
Graduated No
Mouth Wide
ACT Label Yes
Height (Metric) 180 mm
Height (English) 7 in.
Closure Liner Unlined
Closure Material Polypropylene
Closure Size 53 mm
Diameter (Metric) 124.5 mm
Closure Type Screw Cap
Through the Closure Dispensing No
Capacity (English) 32 oz.
Shape Rectangular
Certifications/Compliance USP Class VI; FDA - 21 CFR
Product Type Wide Mouth Bottle
Show More Show Less

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.