Learn More
Thermo Scientific™ Nalgene™ Rectangular HDPE Bottles with Closure

Transport, sample or store liquids and dry materials with these space-efficient bottles featuring excellent chemical resistance to most acids, bases and alcohols.
Supplier: Thermo Scientific™ 20070016
Description
Transport, sample or store liquids and dry materials with these space-efficient bottles that feature excellent chemical resistance to most acids, bases and alcohols. Thermo Scientific™ Nalgene™ Rectangular HDPE Bottles with Closure take up less shelf space than traditional round bottles. HDPE bottles have a sturdy wall construction with straight sides and a wide mouth for easy filling. Made only from high-quality, laboratory-grade, plastic materials for dependably low leachables and extractables. This product is ACT Labeled by My Green Lab to enable data-based sustainable product purchasing decisions.
Features:- Excellent chemical resistance to most acids, bases and alcohols make these excellent containers to protect your valuable samples
- Rectangular shape makes it easy to fit in freezer for storage up to -100°C (-148°F)
- Linerless polypropylene screw closures provide the ultimate in leakproof† protection without the use of a liner that can wrinkle, cause leaks, or contaminate your reagents
- Leakproof†
Includes:
Polypropylene screw caps
Recycling Information: Nalgene reusable HDPE bottles are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually. Nalgene HDPE bottles and their packaging are readily recyclable. Please recycle the corrugate cartons and LDPE bags in which your Nalgene bottles products are packaged. And at end of product life, please wash and recycle your Nalgene HDPE bottles.
Chemical Identifiers
| Translucent | |
| HDPE | |
| Natural, translucent | |
| Yes | |
| Wide | |
| Yes | |
| Unlined | |
| Screw Cap |
| Wide-Mouth Rectangular Bottle | |
| Non-sterile | |
| USP Class VI; FDA - 21 CFR | |
| No | |
| Rectangular | |
| Wide Mouth Bottle | |
| Polypropylene | |
| No |
Specifications
| 16 oz., 500 mL | |
| 16 oz. | |
| Wide-Mouth Rectangular Bottle | |
| 37 mm | |
| 94 mm | |
| 12 | |
| 60 mm | |
| USP Class VI; FDA - 21 CFR | |
| No | |
| Rectangular | |
| 147 mm | |
| 5.8 in. | |
| Polypropylene | |
| 96.5 mm | |
| No |
| 500 mL | |
| Translucent | |
| 1.4 in. | |
| HDPE | |
| 48 | |
| Non-sterile | |
| Natural, translucent | |
| Yes | |
| Wide | |
| Yes | |
| Wide Mouth Bottle | |
| Unlined | |
| 48 mm | |
| Screw Cap |
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.