Learn More
Thermo Scientific™ Nalgene™ Rectangular Amber HDPE Bottles
Protect light-sensitive liquids or dry materials from exposure with these space-efficient, amber HDPE bottles, featuring sturdy walls and a wide mouth for easy filling.
Supplier: Thermo Scientific™ 20090032
Description
Protect light-sensitive liquids or dry materials from exposure. Thermo Scientific™ Nalgene™ Rectangular Amber HDPE Bottles take up less shelf space than traditional round bottles. These high-density, polyethylene bottles have a sturdy wall construction with straight sides and a wide mouth for easy filling. Excellent chemical resistance to most acids, bases and alcohols makes this container great for sampling, shipping or storing liquids or dry materials that are sensitive to UV light.
- Amber polypropylene screw closure
- Amber HDPE reduces UV light transmissions to protect light-sensitive liquids
- Meets U.S. Pharmacopoeia 23 light transmission requirements
- Rectangular shape is great for space-saving use in the freezer to -100°C (-148°F)
- Excellent chemical resistance to most acids, bases and alcohols make these excellent containers to protect your valuable samples
- Linerless caps provide the ultimate in leakproof† protection without the use of a liner that can wrinkle, cause leaks or contaminate your reagents
- Leakproof†
Specifications
| 32 oz., 1000 mL | |
| 32 oz. | |
| Amber Wide-Mouth Rectangular Bottle | |
| 44 mm | |
| 125 mm | |
| 6 | |
| 71 mm | |
| Meet U.S. Pharmacopeia XXIII requirements for maximum light transmission. | |
| No | |
| Rectangular | |
| Wide Mouth Bottle | |
| Unlined | |
| 53 mm | |
| Screw Cap |
| 1 L | |
| Amber | |
| 1.7 in. | |
| HDPE | |
| 24 | |
| Non-sterile | |
| Opaque Amber | |
| Yes | |
| Wide | |
| 180 mm | |
| 7 in. | |
| Polypropylene | |
| 124.5 mm | |
| No |
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.