missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Rapid-Flow™ Sterile Disposable Bottle Top Filters with PES, CN, SFCA or Nylon Membranes
GREENER_CHOICE

Catalog No. 0974109
Change view
Click to view available options
Volume (Metric):
150 mL
500 mL
1000 mL
Material (Membrane):
PES
SFCA
Pore Size:
0.2 μm
0.45 μm
Fits Bottle Neck Size (Metric):
33mm
45mm
20 product options available for selection
Product selection table with 20 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Material (Membrane) Pore Size Fits Bottle Neck Size (Metric)
0974109 150 mL PES 0.2 μm 45mm
0974108 150 mL PES 0.2 μm 33mm
0974062C 150 mL PES 0.45 μm 33mm
0974062D 150 mL PES 0.45 μm 45mm
0974106 500 mL PES 0.2 μm 33mm
0974107 500 mL PES 0.2 μm 45mm
0974064A 500 mL PES 0.45 μm 33mm
0974064B 500 mL PES 0.45 μm 45mm
0974110 1000 mL PES 0.2 μm 33mm
0974111 1000 mL PES 0.2 μm 45mm
0974022D 150 mL SFCA 0.2 μm 33mm
0974022F 150 mL SFCA 0.2 μm 45mm
0974022E 150 mL SFCA 0.45 μm 33mm
0974022G 150 mL SFCA 0.45 μm 45mm
0974022H 500 mL SFCA 0.2 μm 33mm
0974022K 500 mL SFCA 0.2 μm 45mm
0974022J 500 mL SFCA 0.45 μm 33mm
0974022L 500 mL SFCA 0.45 μm 45mm
0974047A 1000 mL SFCA 0.2 μm 33mm
0974047C 1000 mL SFCA 0.2 μm 45mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 20 options available.
20 options
CAUTION: Use only sterile bottles designed for use with vacuum. Always use safety shield during vacuum procedures.
Catalog No. 0974109 Supplier Thermo Scientific™ Supplier No. 5964520
CAUTION: Use only sterile bottles designed for use with vacuum. Always use safety shield during vacuum procedures.

Achieve fast flow rates and high throughput for increased filtration efficiency with Nalgene Rapid-Flow sterile disposable bottle top filters.

Increase filtration efficiency. Thermo Scientific™ Nalgene™ Rapid-Flow™ single use bottle tops feature the exclusive Rapid-Flow support column design, providing fast flow rates and high throughput. The last line of defense against cell culture contamination—Nalgene Rapid-Flow bottle tops are the best choice for the filtration of cell culture media, serum, additives and buffers.

  • Available in PES and SFCA membranes. Devices are color coded according to membrane.
  • Devices feature the fastest flow rate and lowest clogging of comparable alternatives
  • 0.1um PES available for mycoplasma protection
  • 0.2um PES devices are stem cell and mouse embryonic assay (MEA) tested
  • Non-pyrogenic and endotoxin free - Rapid-Flow filters meet the most rigorous testing standards for any vacuum filtration unit
  • Available with Fit for Purpose Sterile documentation

Specifications

Disposable Single-use
Filter Type Membrane Filter
Material Polystyrene
Material (Membrane) PES
Pore Size 0.2 μm
Sterility Sterile
Fits Bottle Neck Size (Metric) 45mm
No. per Case 12
No. per Pack 12
Type Bottle Top Filter
Volume (Metric) 150 mL
Diameter (Metric) 50 mm
Diameter (Metric) Membrane 50 mm
Fit for Purpose Sterile
Show More Show Less