missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Nalgene™ Rapid-Flow™ Sterile Disposable Bottle Top Filters with PES, CN, SFCA or Nylon Membranes

Description
Increase filtration efficiency. Thermo Scientific™ Nalgene™ Rapid-Flow™ single use bottle tops feature the exclusive Rapid-Flow support column design, providing fast flow rates and high throughput. The last line of defense against cell culture contamination—Nalgene Rapid-Flow bottle tops are the best choice for the filtration of cell culture media, serum, additives and buffers.
- Available in PES and SFCA membranes. Devices are color coded according to membrane.
- Devices feature the fastest flow rate and lowest clogging of comparable alternatives
- 0.1um PES available for mycoplasma protection
- 0.2um PES devices are stem cell and mouse embryonic assay (MEA) tested
- Non-pyrogenic and endotoxin free - Rapid-Flow filters meet the most rigorous testing standards for any vacuum filtration unit
- Available with Fit for Purpose Sterile documentation
Specifications
Specifications
| Disposable | Single-use |
| Filter Type | Membrane Filter |
| Material | Polystyrene |
| Material (Membrane) | SFCA |
| Pore Size | 0.2 μm |
| Sterility | Sterile |
| Fits Bottle Neck Size (Metric) | 45mm |
| No. per Case | 12 |
| No. per Pack | 12 |
| Type | Bottle Top Filter |
| Show More |