missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Autoclavable PPCO Wash Bottles
GREENER_CHOICE

Catalog No. 0340914c
Change view
Click to view available options
Capacity (Metric):
500 mL
1000 mL
Closure Size:
28 mm
38-430 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Closure Size
0340914C 1000 mL 38-430 mm
0340914A 500 mL 28 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 0340914C Supplier Thermo Scientific™ Supplier No. 24051000
Get a Water Bottle with Plastics Purchase
View all promotions

Reduce the risk of contamination when dispensing sterile water or reagents by using these autoclavable PPCO wash bottles.

Reduce the risk of contamination when dispensing sterile water or reagents by using autoclavable Thermo Scientific™ Nalgene™ PPCO Wash Bottles. Made from high-quality, laboratory-grade, plastic materials for dependably low leachables and extractables. These squeeze bottles are flexible, contact-clear, and designed to dispense through the top with a closure and stem molded in one piece for reliable service. Bent stem dispensing tube retains its bend after autoclaving but bend angle can be modified by hand while warm if desired. The tight fit of the draw tube and closure prevents separation when squeezing bottle.

  • Polypropylene screw/stem and draw tube
  • Tip can be cut back to provide increased flow

Specifications

Capacity 32 oz., 1000 mL
Capacity (Metric) 1000 mL
Capacity (English) 32 oz.
Color Translucent
Includes Screw/stem and draw tube
Material PPCO
No. per Case 12
No. per Pack 4
Sterility Non-sterile
Closure Included Yes
Graduated No
Mouth Narrow
Shape Cylindrical
Height (Metric) 259 mm
Product Type Wash Bottle
Height (English) 10.2 in.
Closure Material Polypropylene
Closure Size 38-430 mm
Diameter (Metric) 91.4 mm
Closure Type Cap
Through the Closure Dispensing Yes
Autoclavable Autoclavable
Show More Show Less