missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Nalgene™ Polycarbonate, Validation Bottle
For Use With: Perform validation testing; Autoclavable; Closure Material: Polypropylene; Color: White (Translucent)
$697.96 - $2548.36
Specifications
| Autoclavable | Autoclavable |
|---|---|
| Closure Material | Polypropylene |
| For Use With (Application) | Perform validation testing |
| Material | Polycarbonate |
| Mouth Style | Wide |
| Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity | |||||
|
36-104-0070
|
Thermo Scientific™
DS21270030 |
30 mL |
Case of 30 Each for $2,548.36
|
|
|||||
|
02-923-10
|
Thermo Scientific™
DS21270250 |
250 mL |
Case of 6 Each for $697.96
|
|
|||||
|
36-103-1180
|
Thermo Scientific™
DS21272000 |
2 L |
Case of 12 Each for $2,182.64
|
|
|||||
Description
Perform QA lot sample storage of high-valued pharmaceuticals using Thermo Scientific™ Nalgene™ Polycarbonate Validation Bottles. Small-volume bottles are molded of the same materials as larger carboys, but require a lower volume of expensive reagents to perform validation testing.
Autoclavable, transparent polycarbonate construction allows for convenient material compatibility validation.Includes:
White polypropylene screw closure and TPE gasket
Specifications
| Autoclavable | |
| Perform validation testing | |
| Wide | |
| Non-sterile |
| Polypropylene | |
| Polycarbonate | |
| Validation Bottle |