missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Nalgene™ Polycarbonate, Validation Bottle
For Use With: Perform validation testing; Autoclavable; Closure Material: Polypropylene; Color: White (Translucent)
Supplier: Thermo Scientific™ DS21272000
Description
Perform QA lot sample storage of high-valued pharmaceuticals using Thermo Scientific™ Nalgene™ Polycarbonate Validation Bottles. Small-volume bottles are molded of the same materials as larger carboys, but require a lower volume of expensive reagents to perform validation testing.
Autoclavable, transparent polycarbonate construction allows for convenient material compatibility validation.Includes:
White polypropylene screw closure and TPE gasket
Specifications
| Autoclavable | |
| 2 L | |
| 53B | |
| 42 mm | |
| Perform validation testing | |
| Polycarbonate | |
| 12/Cs. | |
| Validation Bottle |
| 0.5 gal. | |
| Polypropylene | |
| Clear | |
| 123 mm | |
| 296 mm | |
| Wide | |
| 12 Pk. | |
| Non-sterile |