missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Petri and Bioassay Dish Racks
GREENER_CHOICE

Catalog No. 0875910A
Change view
Click to view available options
Holds:
Petri Dishes
For Use With (Equipment):
2.36 in. (60mm) dish
3.93 in. (100mm) dish
Autoclavable:
Autoclavable
Not Autoclavable
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Holds For Use With (Equipment) Autoclavable
0875910A Petri Dishes 3.93 in. (100mm) dish Autoclavable
0875910D Petri Dishes 2.36 in. (60mm) dish Not Autoclavable
0875910B Petri Dishes 3.93 in. (100mm) dish Not Autoclavable
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 0875910A Supplier Thermo Scientific™ Supplier No. 59200060

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Secure glass and plastic dishes for safe transport with these petri and bioassay dish racks.

Secure glass and plastic dishes for safe transport with Nalgene™ Petri and Bioassay Dish Racks. Requiring only simple assembly, these clear racks permit side visibility for culture checks during incubation.

  • Clear racks permit side visibility for culture checks during incubation
  • Simple assembly required

Specifications

Autoclavable Autoclavable
Color Clear
Holds Petri Dishes
Material Polycarbonate
For Use With (Equipment) 3.93 in. (100mm) dish
No. per Case 2
No. per Pack 1
Product Type Petri Dish Rack
Diameter (Metric) Inner 100 mm