missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ PETG Diagnostic Bottles with Closure: Sterile
GREENER_CHOICE

Catalog No. 02925100
Change view
Click to view available options
Capacity (Metric):
5 mL
10 mL
20 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
02925100 5 mL
02925101 10 mL
02925102 20 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 02925100 Supplier Thermo Scientific™ Supplier No. 20350005
Only null left

Conduct sterile sampling, storage and shipment of reagents with these clear PETG bottles that are leakproof when used with provided Nalgene lined white HDPE closures.

Conduct sterile sampling, storage and shipment of reagents and buffer solutions using Thermo Scientific™ Nalgene™ PETG Diagnostic Bottles with Closure. These bottles are constructed from non-pyrogenic and non-cytotoxic polyethylene terephthalate copolymer with lined white HDPE (high-density polyethylene) closures.

Features:
  • Sterile to 10-6 SAL
  • Non-pyrogenic and non-cytotoxic
  • 20-415 lined white high-density polyethylene closure attached to PETG vial
  • Leakproof†

Ordering Information:

Serum Vials are packed in shrink-wrapped trays with closures attached.

Specifications

Capacity 0.16 oz., 5 mL
Capacity (Metric) 5 mL
Capacity (English) 0.16 oz.
Diameter (Metric) Inner Neck 11 mm
Material PETG
No. per Case 100
No. per Pack 20
Sterility Sterile
Neck Finish 20-415 mm
Certifications/Compliance Manufactured in compliance with ISO 13485:2016 quality requirements. Serum Vials meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
Closure Included Yes
Mouth Narrow
Height (Metric) 46 mm
Product Type Sample Container
Height (English) 1.81 in.
Diameter (Metric) 22 mm
Through the Closure Dispensing No
Show More Show Less

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes.

Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.