missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ PETG Low Particulate Biotainer Bottles
GREENER_CHOICE

Catalog No. 14388016
Change view
Click to view available options
Capacity (Metric):
125 mL
500 mL
1 L
2 L
5 L
Includes:
Neck Handle
Side Handle
Quantity:
14 x 5 Bottles
20 Bottles
20 x 5 Bottles
4 x 5 Bottles
6 Bottles
7 x 5 Bottles
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Includes Quantity
14388016 5 L Side Handle 6 Bottles
14388011 125 mL Side Handle 20 x 5 Bottles
14388012 500 mL Side Handle 14 x 5 Bottles
14388013 1 L Side Handle 7 x 5 Bottles
14388015 2 L Side Handle 4 x 5 Bottles
14388014 2 L Side Handle 20 Bottles
14388017 5 L Neck Handle 6 Bottles
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 14388016 Supplier Thermo Scientific™ Supplier No. 38341516
Only null left

Nalgene PETG Low Particulate Biotainer Bottles are certified clean to USP <788> standards, making them an excellent solution for biological and pharmaceutical manufacturing.

Specifically designed for pharmaceutical and biological manufacturing, Thermo Scientific™ Nalgene™ Certified Clean PETG Biotainer™ Bottles reduce the risk of carry-over contamination.

  • Specifically designed for pharmaceutical and biological manufacturing
  • Certified Clean, validated to USP <788>
  • <0.5 EU/mL Endotoxin

Specifications

Autoclavable Not Autoclavable
Capacity (Metric) 5 L
Capacity (English) 169.07 fl. oz.
Certifications/Compliance USP <778>
Closure Color White
Closure Liner LDPE
Color Clear
Closure Material Polypropylene
Closure Size 48 mm
Diameter (Metric) Outer Neck 48 mm
For Use With (Application) Media/Buffer Preparation ​, Media/Buffer Storage, Sampling, Storage, Transportation ​, Validation Testing, Drug Substance, Drug Product, Media/Buffer Preparation ​, Media/Buffer Storage, Sampling, Storage, Transportation ​, Validation Testing, Drug Substance, Drug Product
Includes Side Handle
Material PET, PETG
Diameter (Metric) Outer 166 mm
No. per Case 6/Cs.
No. per Pack 1 Ea.
Quantity 6 Bottles
Height (Metric) 299 mm
Sterility Sterile
Length (English) 21.375 in.
Product Type Biotainer, Square Bottle
Width (English) 14.75 in.
Low Particulate USP <788> (Low particulate)
Diameter (Metric) 166 mm
Workflow Step Upstream media/buffer prep and storage. Upstream process solution storage. Downstream validation, downstream intermediate storage and transport. Bulk drug product/substance transportation.
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.