Learn More
Thermo Scientific™ Nalgene™ Non-Phthalate PVC Tubing
GREENER_CHOICE
Lab-grade, autoclavable clear tubing sets the standard for purity and quality, made from highest-grade resins, free from ortho-phthalate plasticizers. Complies with FDA CFR21 for food packaging, USP VI, Cytotoxicity, REACH, Latex-free, RoHS, and California Proposition 65 standards
Supplier:  Thermo Scientific™ 87014140
 View more versions of this product
Catalog No. 1371293
Only null left
Description
Description
Thermo Scientific™ Nalgene™ Non-Phthalate Clear Plastic PVC Tubing is manufactured from the highest-grade resin materials and performs like classic Nalgene 180 tubing but is free from orthophthalate DEHP plasticizers. Complies with FDA CFR21 for food packaging, USP VI, Cytotoxicity, REACH, Latex-free, RoHS, and complies with California proposition 65. Resists hardening and discoloration from a wide range of laboratory chemicals and does not impart odor or taste making this tubing ideal for a wide range of non-medical, general lab, research and pharmaceutical applications.
  • Lab-grade, autoclavable clear tubing sets the standard for purity and quality, made from highest-grade resins and plasticizers
  • Free from ortho phthalate plasticizers
  • Resists hardening and discoloration from a wide range of chemicals - demonstrates dimensional stability
  • Equivalent to Tygon™ E-3603 tubing
  • Marked with I.D. and regulatory compliance as well as at 12 in. intervals for easy identification and measurement
  • Durometer hardness is Shore A, 60
  • Does not impart odor or taste
  • Can write on with Nalgene Lab marker
  • Slips easily over glass and plastic fittings; grips securely
  • Autoclavable/Transparent
Compliance
FDA CFR21 for food packaging, USP VI, Cytotoxicity, REACH, Latex-free, RoHS, and California Proposition 65
Specifications
Chemical Identifiers
Meets FDA CFR21 for food packaging, USP VI, Cytotox, REACH, Latex-free, RoHS, and CA Prop 65
Autoclavable
Non-sterile
Clear
4 x OD bend radius
160°F
Flexible Tubing
Non-Phthalate PVC Tubing
PVC
Autoclave, Gas, Chemical
60 (Shore A)
Very low
71°C
Specifications
Meets FDA CFR21 for food packaging, USP VI, Cytotox, REACH, Latex-free, RoHS, and CA Prop 65
Autoclavable
PVC
100 ft.
1 piece of tubing 100 ft in length
Autoclave, Gas, Chemical
60 (Shore A)
40 psig at 73°F (23°C), 20 psig at 125°F (51°C)
160°F
Flexible Tubing
Non-Phthalate PVC Tubing
3/8 in.
5/8 in.
1/8 in.
Non-sterile
Clear
4 x OD bend radius
Very low
71°C
Product Suggestions
Product Suggestions