Learn More
Thermo Scientific™ Nalgene™ Natural PPCO Diagnostic Bottles with Closure: Bulk Pack

Sample, store and ship small-volume, valuable reagents with these autoclavable bottles that can be used in test kits and many small-volume applications.
Supplier: Thermo Scientific™ 3120069050
Description
Sample, store and ship small-volume, valuable reagents. Thermo Scientific™ Nalgene™ Natural PPCO Diagnostic Bottles with Closure are durable, autoclavable, and used in test kits and many small-volume applications. These natural, translucent polypropylene copolymer bottles feature uniform walls resistant to splitting or puncturing. Linerless closure, included, ensures performance without wrinkles that can cause leaks or contamination of your solutions.
Features:- Nonsterile
- Autoclavable
Ordering Information:
Bottles and closures are packed in separate polybags within the same carton. Packed 2,000 per case.
Chemical Identifiers
| Translucent | |
| 2000 | |
| Non-sterile | |
| Yes | |
| Sample Container |
| PPCO | |
| 2000 | |
| Manufactured in compliance with ISO 13485:2016 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI. | |
| Narrow | |
| Autoclavable |
Specifications
| 0.5 oz., 15 mL | |
| 0.5 oz. | |
| 14 mm | |
| 2000 | |
| Non-sterile | |
| Manufactured in compliance with ISO 13485:2016 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI. | |
| No | |
| Round | |
| Sample Container | |
| Unlined | |
| 20-415 mm | |
| Screw Cap |
| 15 mL | |
| Translucent | |
| PPCO | |
| 2000 | |
| 20-415 mm | |
| Yes | |
| Narrow | |
| 58 mm | |
| 2.3 in. (With Closure); 2.2 in. (Without Closure) | |
| Polypropylene | |
| 25 mm | |
| Autoclavable |
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.