missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Natural HDPE Diagnostic Bottles with Closure: Bulk Pack
GREENER_CHOICE

Sample, store and ship small-volume, valuable reagents in these bottles with polypropylene closures that are leakproof when used with Nalgene closures. They can be used in test kits and many small-volume applications as well.

Supplier:  Thermo Scientific™ 3120029050

 View more versions of this product

Catalog No. 0331325A


Only null left

Description

Description

Sample, store and ship small-volume, valuable reagents. Thermo Scientific™ Nalgene Diagnostic Bottles with closures are used in test kits and many small-volume applications. Natural, translucent high-density polyethylene provides a durable design with uniform walls that are resistant to splitting or puncturing.

  • Nonsterile
  • Natural, translucent high-density polyethylene
  • Natural, linerless polypropylene closures
  • Durable, uniform walls are resistant to splitting or puncturing
  • Leakproof†

Ordering Information:

Bottles and closures are packed in separate polybags within the same carton. 2000 per case. † The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Specifications

Chemical Identifiers

HDPE
2000
Manufactured in compliance with ISO 13485:2016 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
Narrow
Unlined
2000
Non-sterile
Yes
Sample Container
No

Specifications

0.5 oz., 15 mL
0.5 oz.
14 mm
2000
Non-sterile
Manufactured in compliance with ISO 13485:2016 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
No
Round
Sample Container
2.3 in.
Polypropylene
25 mm
No
15 mL
Natural
HDPE
2000
20-415 mm
Yes
Narrow
58 mm
0.98 in.
Unlined
20-415 mm
Screw Cap
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.