Learn More
Thermo Scientific™ Nalgene™ Natural HDPE Diagnostic Bottles with Closure: Bulk Pack

Sample, store and ship small-volume, valuable reagents in these bottles with polypropylene closures that are leakproof when used with Nalgene closures. They can be used in test kits and many small-volume applications as well.
Supplier: Thermo Scientific™ 3120029125
Description
Sample, store and ship small-volume, valuable reagents. Thermo Scientific™ Nalgene Diagnostic Bottles with closures are used in test kits and many small-volume applications. Natural, translucent high-density polyethylene provides a durable design with uniform walls that are resistant to splitting or puncturing.
- Nonsterile
- Natural, translucent high-density polyethylene
- Natural, linerless polypropylene closures
- Durable, uniform walls are resistant to splitting or puncturing
- Leakproof†
Ordering Information:
Bottles and closures are packed in separate polybags within the same carton. 2000 per case. † The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Chemical Identifiers
| HDPE | |
| 2000 | |
| Manufactured in compliance with ISO 13485:2016 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI. | |
| Narrow | |
| Unlined |
| 2000 | |
| Non-sterile | |
| Yes | |
| Sample Container | |
| No |
Specifications
| 0.125 oz., 4 mL | |
| 0.125 oz. | |
| 8 mm | |
| 2000 | |
| Non-sterile | |
| Manufactured in compliance with ISO 13485:2016 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI. | |
| Narrow | |
| Sample Container | |
| Unlined | |
| No |
| 4 mL | |
| Translucent | |
| HDPE | |
| 2000 | |
| 13-415 mm | |
| Yes | |
| 41 mm | |
| 1.61 in. | |
| 16 mm |
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.