missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Narrow-Mouth HDPE Lab Quality Bottles with Closure
GREENER_CHOICE

Catalog No. 029236A
Change view
Click to view available options
Capacity (Metric):
4 mL
8 mL
15 mL
30 mL
60 mL
125 mL
175 mL
250 mL
500 mL
1 L
Closure Size:
13 mm
20 mm
24 mm
28 mm
38-430 mm
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Closure Size
029236A 4 mL 13 mm
0292311A 8 mL 20 mm
029238A 15 mL 20 mm
02923A 30 mL 20 mm
02923B 60 mL 20 mm
02923C 125 mL 24 mm
02923CC 175 mL 24 mm
02923D 250 mL 24 mm
0292311F 500 mL 38-430 mm
02923E 500 mL 28 mm
02923F 1 L 38-430 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Catalog No. 029236A Supplier Thermo Scientific™ Supplier No. 20029125

Store, ship and package liquids with these highly reliable, lab-quality bottles, specifically designed for long-term demanding lab use.

Store, ship, and package liquids using highly reliable and durable Thermo Scientific™ Nalgene™ Narrow-Mouth HDPE Lab Quality Bottles with Closure. These semi-rigid, translucent, all-purpose bottles are specifically designed for long-term, demanding lab use and offer excellent chemical resistance. Made only from high quality, laboratory-grade plastic materials for dependably low leachables and extractables. This product is ACT Labeled by My Green Lab to enable data-based sustainable product purchasing decisions.

  • Versatile and indispensable in the lab
  • Uniform walls provide durability and resistance to splitting or punctures
  • Can be used in a freezer up to -100°C (-149°F)
  • Linerless polypropylene closures provide the ultimate in leakproof† protection without the use of liner that can wrinkle, cause leaks or contaminate your reagents
  • Participates in the ACT Label Program
  • Leakproof†

Specifications

Capacity 0.125 oz., 4 mL
Capacity (Metric) 4 mL
Capacity (English) 0.125 oz.
Color Translucent
Description Lab Quality Narrow-Mouth Bottle
Diameter (Metric) Inner Neck 8 mm
Material HDPE
No. per Case 72
No. per Pack 12
Sterility Non-sterile
Closure Color Natural
Neck Finish 13 mm
Certifications/Compliance Manufactured in compliance with ISO 13485:2016 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
Closure Included Yes
Graduated No
Mouth Narrow
Shape Round
ACT Label Yes
Height (Metric) 41 mm
Product Type Storage Bottle
Width (English) 0.62 in.
Height (English) 1.6 in.
Closure Liner Unlined
Closure Material Polypropylene
Closure Size 13 mm
Diameter (Metric) 16 mm
Closure Type Screw Cap
Through the Closure Dispensing No
Show More Show Less

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.