Learn More
Thermo Scientific™ Nalgene™ Narrow-Mouth HDPE IP2 Bottles
GREENER_CHOICE
Use these shipping bottles when designing, assembling and certifying combination packaging. They resist splitting and puncturing from shipping shocks.
$84.21 - $198.34
Specifications
Color Translucent
Description Narrow-Mouth Passport IP2 Bottles
Includes Polypropylene (PP) screw caps
Material HDPE
Certifications/Compliance Meets IATA Dangerous Goods Regulations Section 5.0.2.9.
View More Specs
Products 7
Catalog Number Mfr. No. Capacity (Metric) Closure Size Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Closure Size Price Quantity & Availability  
033126
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
20990001
30 mL 20-415 mm
Pack of 12 Each for $84.21
Case of 6 Pack for $505.26
Only null left
033127
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
20990002
60 mL 20-415 mm
Pack of 12 Each for $89.34
Case of 6 Pack for $536.04
Only null left
033128
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
20990004
125 mL 24-415 mm
Pack of 12 Each for $121.79
Case of 6 Pack for $730.74
Only null left
033129
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
20990008
250 mL 24-415 mm
Pack of 12 Each for $187.76
Case of 6 Pack for $1,126.56
Only null left
0331210
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
20990016
500 mL 28-415 mm
Pack of 12 Each for $198.34
Case of 4 Pack for $793.36
Only null left
0331211
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
20990032
1000 mL 38-430 mm
Pack of 6 Each for $195.21
Case of 4 Pack for $780.84
Only null left
0331212
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
20990010
4000 mL 38-430 mm
Each for $138.97
Case of 6 Each for $833.82
Only null left
Description
Description
Utilize Thermo Scientific™ Nalgene™ Narrow-Mouth HDPE IP2 Bottles as inner containers for designing, assembling and certifying combination packaging. These high density polyethylene bottles are safe, economical and designed to resist splitting and puncturing from shipping shocks.
  • Heavy-duty walls resist splitting and puncturing
  • Leakproof† when used with Nalgene closures
Includes:
Polypropylene screw caps
Specifications
Specifications
Translucent
Polypropylene (PP) screw caps
Meets IATA Dangerous Goods Regulations Section 5.0.2.9.
Narrow
IP2 Bottle
Screw Cap
Narrow-Mouth Passport IP2 Bottles
HDPE
Yes
Round
Polypropylene
Videos
SDS
Documents
Documents
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.