Learn More
Thermo Scientific™ Nalgene™ Narrow-Mouth HDPE IP2 Bottles with Closure: Bulk Pack

Ship and store liquid reagents using these high density, durable polyethylene bottles with closure. Recommended for customers who design, assemble and certify their own combination packaging.
Supplier: Thermo Scientific™ 3120990008
Description
Ship and store liquid reagents using durable Thermo Scientific™ Nalgene™ Narrow-Mouth HDPE IP2 Bottles with Closure. These high density polyethylene bottles are a safe, economical solution for customers who design, assemble and certify their own combination packaging.
Durable bottles for shipping liquid reagents
- HDPE bottle with PP closure, bulk pack
- Narrow mouth
- Complies with IATA and ICAO regulations
Ordering Information:
Bottles and closures are bulk packed in separate polybags within the same carton
Recommended for:
For storage and shipment of liquid reagents.
Chemical Identifiers
| Translucent | |
| Storage and Shipment of Liquid Reagents | |
| Manufactured in compliance with ISO 13485:2016 quality requirements. Bottles and closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI. Bottles are evaluated at 15psi (103kPa) per 49 CFR 173.27 (c)(2), ICAO Technical Instructions Part 4; 1.1.6, and IATA Dangerous Goods Regulations Section 5.0.2.9. | |
| Narrow | |
| IP2 Bottle |
| IP2 Narrow-Mouth Bottles | |
| HDPE | |
| Yes | |
| Round | |
| Polypropylene |
Specifications
| 8 oz., 250 mL | |
| 8 oz. | |
| IP2 Narrow-Mouth Bottles | |
| 18 mm | |
| 133 mm | |
| 250 | |
| Manufactured in compliance with ISO 13485:2016 quality requirements. Bottles and closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI. Bottles are evaluated at 15psi (103kPa) per 49 CFR 173.27 (c)(2), ICAO Technical Instructions Part 4; 1.1.6, and IATA Dangerous Goods Regulations Section 5.0.2.9. | |
| Narrow | |
| 133 mm | |
| 5.2 in. | |
| 24-415 mm |
| 250 mL | |
| Translucent | |
| 18 mm | |
| Storage and Shipment of Liquid Reagents | |
| HDPE | |
| 250 | |
| Yes | |
| Round | |
| IP2 Bottle | |
| Polypropylene | |
| 61 mm |