Learn More
Thermo Scientific™ Nalgene™ Narrow-Mouth Amber HDPE Lab Quality Bottles

Store, ship and package light-sensitive liquids using these narrow-mouth, amber HDPE bottles that offer excellent chemical resistance to most acids, bases and alcohols.
$70.16 - $272.41
Specifications
| Color | Opaque Amber |
|---|---|
| Description | Lab Quality Amber Narrow-Mouth Bottle |
| Material | HDPE |
| Sterility | Non-sterile |
| Closure Color | Amber |
| Catalog Number | Mfr. No. | Capacity (Metric) | Closure Size | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity (Metric) | Closure Size | Price | Quantity & Availability | ||||
029238F
![]() |
Thermo Scientific™
20049125 |
4 mL | 13 mm |
Pack of 12 Each for $73.86
Case of 6 Pack for $443.16
|
|
||||
0292311C
![]() |
Thermo Scientific™
20049025 |
8 mL | 20 mm |
Pack of 12 Each for $70.16
Case of 6 Pack for $420.96
|
|
||||
029238E
![]() |
Thermo Scientific™
20049050 |
15 mL | 20 mm |
Pack of 12 Each for $83.28
Case of 6 Pack for $499.68
|
|
||||
029235A
![]() |
Thermo Scientific™
20040001 |
30 mL | 20 mm |
Pack of 12 Each for $106.79
Case of 6 Pack for $640.74
|
|
||||
029235B
![]() |
Thermo Scientific™
20040002 |
60 mL | 20 mm |
Pack of 12 Each for $120.76
Case of 6 Pack for $724.56
|
|
||||
029235C
![]() |
Thermo Scientific™
20040004 |
125 mL | 24 mm |
Pack of 12 Each for $136.45
Case of 6 Pack for $818.70
|
|
||||
029235D
![]() |
Thermo Scientific™
20040008 |
250 mL | 24 mm |
Pack of 12 Each for $183.38
Case of 6 Pack for $1,100.28
|
|
||||
029235E
![]() |
Thermo Scientific™
20040016 |
500 mL | 28 mm |
Pack of 12 Each for $272.41
Case of 4 Pack for $1,089.64
|
|
||||
029235F
![]() |
Thermo Scientific™
20040032 |
1 L | 38-430 mm |
Pack of 6 Each for $229.34
Case of 4 Pack for $917.36
|
|
||||
Description
Store, ship and package light-sensitive liquids using Thermo Scientific™ Nalgene™ Narrow-Mouth Amber HDPE Lab Quality Bottles. Featuring excellent chemical resistance to most acids, bases and alcohols, these high-density polyethylene bottles are leakproof† when used with appropriate Nalgene closures. Made only from high-quality, laboratory-grade plastic materials for dependably low leachables and extractables.
Features:- Opaque amber reduces UV light transmissions to protect light-sensitive liquids
- Meets U.S. Pharmacopoeia 23 requirements for light transmission
- Versatile bottles where exposure to light needs to be restricted
- Can be used in freezer to -100°C (-148°F) for long term storage
- Linerless caps provide the ultimate in leakproof† protection without the use of a liner that can wrinkle, cause leaks or contaiminate your reagents
- Leakproof† when used with Nalgene closures
Includes:
Amber polypropylene screw caps
Note:
Nalgene LDPE, HDPE, PP and PMP containers can be used for long-term storage, but direct UV exposure should be avoided.
Recycling Information:
Nalgene reusable HDPE bottles are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually. Nalgene HDPE bottles and their packaging are readily recyclable. Please recycle the corrugate cartons and LDPE bags in which your Nalgene bottles products are packaged. And at end of product life, please wash and recycle your Nalgene HDPE bottles.
Specifications
| Opaque Amber | |
| HDPE | |
| Amber | |
| Yes | |
| Round | |
| Polypropylene | |
| No |
| Lab Quality Amber Narrow-Mouth Bottle | |
| Non-sterile | |
| Meet U.S. Pharmacopeia XXIII requirements for maximum light transmission. | |
| Narrow | |
| Unlined | |
| Screw Cap |
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.