missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Narrow-Mouth Amber HDPE Lab Quality Bottles

Store, ship and package light-sensitive liquids using these narrow-mouth, amber HDPE bottles that offer excellent chemical resistance to most acids, bases and alcohols.

Supplier:  Thermo Scientific™ 20049050

 View more versions of this product

Catalog No. 02-923-8E



Only null left

Description

Description

Store, ship and package light-sensitive liquids using Thermo Scientific™ Nalgene™ Narrow-Mouth Amber HDPE Lab Quality Bottles. Featuring excellent chemical resistance to most acids, bases and alcohols, these high-density polyethylene bottles are leakproof† when used with appropriate Nalgene closures. Made only from high-quality, laboratory-grade plastic materials for dependably low leachables and extractables.

Features:

  • Opaque amber reduces UV light transmissions to protect light-sensitive liquids
  • Meets U.S. Pharmacopoeia 23 requirements for light transmission
  • Versatile bottles where exposure to light needs to be restricted
  • Can be used in freezer to -100°C (-148°F) for long-term storage
  • Linerless caps provide the ultimate in leakproof† protection without the use of a liner that can wrinkle, cause leaks or contaiminate your reagents

Includes:

Amber polypropylene screw caps

Note:

Nalgene LDPE, HDPE, PP and PMP containers can be used for long-term storage, but direct UV exposure should be avoided.

Specifications

Specifications

0.5 oz., 15 mL
0.5 oz.
Lab Quality Amber Narrow-Mouth Bottle
HDPE
12
Amber
Meet U.S. Pharmacopeia XXIII requirements for maximum light transmission.
No
Round
Storage Bottle
2.3 in.
Polypropylene
25 mm
No
15 mL
Opaque Amber
14 mm
72
Non-sterile
20 mm
Yes
Narrow
58 mm
0.98 in.
Unlined
20 mm
Screw Cap
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.