Learn More
Thermo Scientific™ Nalgene™ Large Narrow-Mouth LDPE Bottles

Collect and store distilled water in bottles that offer flexibility, impact resistance and visibility of contents.
$77.17 - $181.59
Specifications
| Color | Translucent |
|---|---|
| Description | Large Narrow-Mouth Bottle |
| Material | LDPE |
| No. per Case | 6 |
| No. per Pack | 1 |
| Catalog Number | Mfr. No. | Capacity (Metric) | Closure Size | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity (Metric) | Closure Size | Price | Quantity & Availability | ||||
02923G
![]() |
Thermo Scientific™
22020005 |
2 L | 38-430 mm |
Each for $77.17
Case of 6 Each for $463.02
|
|
||||
02923J
![]() |
Thermo Scientific™
22020010 |
4 L | 38-430 mm |
Each for $106.79
Case of 6 Each for $640.74
|
|
||||
02923K
![]() |
Thermo Scientific™
22020020 |
8 L | 53B |
Each for $181.59
Case of 6 Each for $1,089.54
|
|
||||
Description
Store chemicals and standards and collect and store distilled water. Thermo Scientific™ Nalgene™ Large Narrow-Mouth LDPE Bottles are translucent and offer flexibility, impact resistance and visibility of contents. The extremely low trace metal content in low-density polyethylene makes them ideal for trace metal analysis. These bottles provide excellent chemical resistance to most acids, bases and alcohols. Made only from high-quality, laboratory-grade, plastic materials for dependably low leachables and extractables.
Features:- Excellent chemical resistance to most acids, bases and alcohols make these versatile bottles
- Can be used in freezer to -100°C (-148°F) for long-term storage
- 4 and 8L sizes feature a built-in shoulder loop on larger sizes for attaching identification tag
- Translucence allows for easy viewing of liquid levels
- Linerless caps ensure leakproof† protection without the use of liner that can wrinkle, cause leaks or contiminate your reagents
- Leakproof† when used with Nalgene closures
- Bottle and Carton are readily recyclable in most communities
- Manufactured from non-toxic materials
Includes:
Polypropylene screw caps, leakproof† when used with Nalgene bottles with the matching finish
Note:
Nalgene LDPE, HDPE, PP and PMP containers can be used for long-term storage, but direct UV exposure should be avoided
.
Recycling Information:
Nalgene reusable LDPE bottles are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually. Nalgene LDPE bottles and their packaging are readily recyclable. Please recycle the corrugate cartons and LDPE bags in which your Nalgene bottles products are packaged. And at end of product life, please wash and recycle your Nalgene LDPE bottles.
Specifications
| Translucent | |
| LDPE | |
| 1 | |
| White | |
| Yes | |
| Round | |
| Unlined | |
| No |
| Large Narrow-Mouth Bottle | |
| 6 | |
| Non-sterile | |
| Manufactured in compliance with ISO 13485:2016 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI. | |
| Narrow | |
| Narrow Mouth Bottle | |
| Polypropylene |
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.