missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Large Narrow-Mouth Amber Bottles
GREENER_CHOICE

Catalog No. 028846A
Change view
Click to view available options
Capacity (Metric):
2 L
4 L
8 L
Closure Size:
53B
38-430 mm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Closure Size
028846A 2 L 38-430 mm
028846B 4 L 38-430 mm
028846D 8 L 53B
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 028846A Supplier Thermo Scientific™ Supplier No. 22040005
Only null left

Protect light-sensitive liquids with these amber bottles that reduce UV light transmissions and feature excellent chemical resistance to most acids, bases and alcohols.

Protect light-sensitive liquids with Thermo Scientific™ Nalgene™ Large Amber Narrow-Mouth Bottles, which reduce UV light transmissions. Useful for storing chemicals and standards, as well as for collecting and storing distilled water, these bottles display excellent chemical resistance to most acids, bases and alcohols.

Features:

  • Opaque amber reduces UV light transmissions to protect light-sensitive liquids
  • Meet the U.S. Pharmacopoeia 23 requirements for light transmission
  • Excellent chemical resistance to most acids, bases and alcohols make these versatile bottles where exposure to light needs to be restricted
  • 4 and 8L sizes feature a built-in shoulder loop on larger sizes for attaching identification tag
  • Linerless caps provide the ultimate in leakproof† protection without the use of a liner that can wrinkle, cause leaks, or contaminate your reagents
  • Autoclavable/Leakproof†

Note:

Completely disengage threads or remove cap before autoclaving. Nalgene LDPE, HDPE, PP and PMP containers can be used for long-term storage, but direct UV exposure should be avoided.

Specifications

Capacity 0.5 gal., 2 L
Capacity (Metric) 2 L
Capacity (English) 0.5 gal.
Color Amber
Description Amber Narrow-Mouth Bottle
Diameter (Metric) Inner Neck 25 mm
Material PPCO
No. per Case 6
No. per Pack 1
Sterility Non-sterile
Closure Color Amber
Neck Finish 38 mm
Certifications/Compliance Comply with U.S. Pharmacopeia XXIII requirements for maximum allowable light transmission
Closure Included Yes
Graduated No
Mouth Narrow
Shape Round
Height (Metric) 249 mm
Product Type Narrow Mouth Bottle
Width (English) 4.75 in.
Height (English) 9.8 in.
Closure Liner Unlined
Closure Material Polypropylene
Closure Size 38-430 mm
Diameter (Metric) 121 mm
Closure Type Polypropylene Closure
Through the Closure Dispensing No
Autoclavable Autoclavable
Show More Show Less

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.