missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Large Cylindrical HDPE Containers with Covers
GREENER_CHOICE

Store and transport dry and solid materials, specimens and general lab supplies in these stackable containers which feature positive-lock lids that form a tight seal.

Supplier:  Thermo Scientific™ 71420015

 View more versions of this product

Catalog No. 11843H



Only null left

Description

Description

Store and transport dry and solid materials, specimens and general lab supplies using Thermo Scientific™ Nalgene™ Large Cylindrical HDPE Containers with Covers. Containers are stackable and have positive-lock lids that seal tight for storing both dry and solid materials.

Features:
  • High-density, polyethylene containers provide excellent chemical and temperature resistance
  • Easy-grip, molded-in handles are comfortable and durable for lifting and moving
  • Positive-lock lid provides a tight seal for transport or storage
  • NSF approved for food and beverage storage
Specifications

Chemical Identifiers

HDPE
Molded
1 Ea.
NSF approved for food and beverage storage.
Lid

Specifications

Container with Cover
4 gal.
NSF approved for food and beverage storage.
30.5 cm
9 in.
Lid
HDPE
15 L
Container Large Round w/Cover HDPE 15 L
Molded
24 cm
1 Ea.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions