Learn More
Thermo Scientific™ Nalgene™ Griffin Low-Form PFA Plastic Beakers
GREENER_CHOICE
Handle temperatures from -270° to +250°C using these sturdy and tough low-form PFA beakers, ideal for continual lab usage.
Supplier:  Thermo Scientific™ 15100250
 View more versions of this product
Catalog No. 0259122C
Only null left
Alert:
Not for use on hot plates.
Description
Description
Handle temperatures from -270° to +250°C using sturdy and tough Thermo Scientific™ Nalgene™ Griffin Low-Form PFA Beakers. Molded from chemically resistant perfluoroalkoxy resin and featuring a heavy-wall design, these beakers are ideal for continual lab usage.
  • No-drip, single-spout design for precision pouring control
  • Highly resistant to most chemicals, including the strongest sulfuric, nitric, chromic and phosphoric acids — will not contaminate contents with leachables
  • Autoclavable/PFA
  • Not for use on hotplates
Specifications
Chemical Identifiers
PFA
Reusable
Griffin, Low Form
Autoclavable
Not Graduated
Specifications
Natural translucent
250 mL
1/Pk.; 6/Cs.
Autoclavable
No
Griffin, Low Form
Beaker Griffin Low-Form PFA 250 mL
PFA
Beaker
Reusable
Not Graduated
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Accessories