Learn More
Thermo Scientific™ Nalgene™ Friction-Fit Polypropylene Closures for Plastic Centrifuge Tubes

Use these friction-fit polypropylene closures featuring a convenient pull tab for easy removal from centrifuge tubes.
Supplier: Thermo Scientific™ DS31110030
Description
Easily remove Thermo Scientific™ Nalgene™ Friction-Fit Polypropylene Closures for Plastic Centrifuge Tubes from the centrifuge with a convenient pull tab. These autoclavable closures are designed to fit Nalgene™ round and conical centrifuge tubes. They may not be liquid-tight during high-speed centrifugation.
Ordering Information:
DS3111-0030 fits non-lipped 50mL centrifuge tubes only.
Chemical Identifiers
| Centrifuge Tube Cap | |
| Designed to be compatible with Nalgene round and conical centrifuge tubes | |
| 20 | |
| Nalgene |
| Autoclavable | |
| Polypropylene | |
| 20 |
Specifications
| 30 mm | |
| Autoclavable | |
| Polypropylene | |
| 20 | |
| Tube with 30 mm O.D. |
| Centrifuge Tube Cap | |
| Designed to be compatible with Nalgene round and conical centrifuge tubes | |
| 20 | |
| Nalgene |