missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Friction-Fit Polypropylene Closures for Plastic Centrifuge Tubes
GREENER_CHOICE

Use these friction-fit polypropylene closures featuring a convenient pull tab for easy removal from centrifuge tubes.

Supplier:  Thermo Scientific™ DS31110017

 View more versions of this product

Catalog No. 0556211F


Only null left

Description

Description

Easily remove Thermo Scientific™ Nalgene™ Friction-Fit Polypropylene Closures for Plastic Centrifuge Tubes from the centrifuge with a convenient pull tab. These autoclavable closures are designed to fit Nalgene™ round and conical centrifuge tubes. They may not be liquid-tight during high-speed centrifugation.

Ordering Information:
DS3111-0030 fits non-lipped 50mL centrifuge tubes only.

Specifications

Chemical Identifiers

Centrifuge Tube Cap
Designed to be compatible with Nalgene round and conical centrifuge tubes
20
Nalgene
Autoclavable
Polypropylene
20

Specifications

17 mm
Autoclavable
Polypropylene
20
Tube with 17 mm O.D.
Centrifuge Tube Cap
Designed to be compatible with Nalgene round and conical centrifuge tubes
20
Nalgene
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions