missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Dropper Bottles

Use for accurate, reliable, repeatable dispensing of various reagents. These dropper bottles are made of opaque LDPE and are ideal for UV lightsensitive solutions. An excellent alternative to pipetting devices.

Supplier:  Thermo Scientific™ 27519125

 View more versions of this product

Catalog No. 03-007-17A


Only null left

Description

Description

Choose Thermo Scientific™ Nalgene™ LDPE White Dropper Bottles for accurate, reliable, repeatable dispensing of various reagents and as an excellent alternative to pipetting devices. Nalgene™ Dropper Bottles are made of opaque LDPE and are ideal for UV lightsensitive solutions. Delivers 40μL drops (based on water; viscosity affects drop size), dispensed one drop at a time. Excellent chemical resistance; suitable for most biotech, diagnostic and pharmaceutical applications.

  • Dropper control tip snaps into place for a secure fit
  • Dispensing tip and closures included, choose from white or multi-colored caps
  • Leakproof†
Specifications

Specifications

4 mL
White
Dispensing tip and closures
25
White
Yes
Round
15-415 mm
Yes
0.13 oz.
LDPE Dropper Bottle
LDPE
25
ISO 13485:2003
40 μL
Dropping Bottle
Dropper Control Tip
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.