Learn More
Thermo Scientific™ Nalgene™ Dropper Bottles with Control Dispensing Tip

Repeatedly dispense reagents accurately using these dropper bottles, an excellent alternative to pipetting devices.
Supplier: Thermo Scientific™ 27529025
Description
Repeatedly dispense reagents accurately using Thermo Scientific™ Nalgene™ Dropper Bottles with Control Dispensing Tip. An excellent alternative to pipetting devices, these dropper bottles deliver 40μL drops (based on water; viscosity affects drop size), dispensed one drop at a time. Featuring excellent chemical resistance, these specialty bottles are suitable for most biotech, diagnostic and pharmaceutical applications.
Features:- Contact-clear, natural, low-density polyethylene bottle for viewing content
- Dropper control tip snaps into place for a secure fit
- Dispensing tip and closures included; chose white or multi-colored closures
- Leakproof†/Transparent
Ordering Information:
This product comes in a case of 25.
Chemical Identifiers
| Translucent | |
| 25 | |
| 40 μL | |
| Dropping Bottle | |
| Yes |
| LDPE | |
| 25 | |
| Round | |
| 15-415 mm |
Specifications
| 8 mL | |
| Translucent | |
| 25 | |
| Assorted | |
| Yes | |
| 40 μL | |
| 64.6 mm | |
| 0.78 in. | |
| 15-415 mm | |
| Yes |
| 0.27 oz. | |
| LDPE | |
| 25 | |
| ISO 13485:2003 | |
| 8 mL | |
| Round | |
| Dropping Bottle | |
| 2.54 in. | |
| 19.7 mm |
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.