missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Dropper Bottles with Control Dispensing Tip
GREENER_CHOICE

Repeatedly dispense reagents accurately using these dropper bottles, an excellent alternative to pipetting devices.

Supplier:  Thermo Scientific™ 27509125

 View more versions of this product

Catalog No. 0300716A


Only null left

Description

Description

Repeatedly dispense reagents accurately using Thermo Scientific™ Nalgene™ Dropper Bottles with Control Dispensing Tip. An excellent alternative to pipetting devices, these dropper bottles deliver 40μL drops (based on water; viscosity affects drop size), dispensed one drop at a time. Featuring excellent chemical resistance, these specialty bottles are suitable for most biotech, diagnostic and pharmaceutical applications.

Features:
  • Contact-clear, natural, low-density polyethylene bottle for viewing content
  • Dropper control tip snaps into place for a secure fit
  • Dispensing tip and closures included; chose white or multi-colored closures
  • Leakproof†/Transparent

Ordering Information:

This product comes in a case of 25.

Specifications

Chemical Identifiers

Translucent
25
40 μL
Dropping Bottle
Yes
LDPE
25
Round
15-415 mm

Specifications

4 mL
Translucent
25
White
4 mL
Round
Dropping Bottle
2.12 in.
19.7 mm
0.13 oz.
LDPE
25
ISO 13485:2003
40 μL
53.8 mm
0.78 in.
15-415 mm
Yes
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.