missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Nalgene™ Disposable Polypropylene Robotic Reservoirs
Description
Increase walk-away time and assay efficiency with Thermo Scientific™ Nalgene™ Disposable Polypropylene Robotic Reservoirs. Featuring a standard microplate format with 300mL volume, the convoluted design minimizes dead volume and has baffles to reduce splashing. The flat bottom version is designed to meet a variety of assay needs in both manual and automated environments.
- Convoluted bottom design is most effective with a 96-well format.
- Flat Bottom design can be used with single, six-, 12-, 96-, 384- and 1536-well formats.
- Polypropylene construction withstands robotic liquid handling, making it easier to run automated assays and increasing walk-away time.
- Reservoir is compatible with most robotic arms, plate stackers (hotels), plate cranes, and platform configurations.
- Reservoir volume is 300mL (345mL brim capacity), with molded-in graduations every 100mL.
- Reservoir is nestable for easy automated stacking and handling.
Specifications
Specifications
| Certifications/Compliance | American National Standards (ANSI) (ANSI/SBS 1-2004) |
| Color | Natural |
| Capacity (Metric) | 300 mL |
| Dimensions (L x W x H) | 3.4 x 5 x 1.75 in. (86 x 128 x 43 mm) |
| Material | Polypropylene |
| Sterility | Non-sterile |
| Type | Reservoir |
| Height (English) | 1.75 in. |
| Height (Metric) | 43 mm |
| Length (English) | 3.4 in. |
| Show More |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.