missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Clear PETG Diagnostic Bottles with Closure: Sterile, Bulk Pack
GREENER_CHOICE

Catalog No. p-4521862
Change view
Click to view available options
Capacity (Metric):
5 mL
10 mL
20 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
02925103 5 mL
02925104 10 mL
02925105 20 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 02925103 Supplier Thermo Scientific™ Supplier No. 3420350005
Only null left

Sample, store and ship small-volume, valuable reagents with clear PETG diagnostic bottles that are ideal for use in many sterile, small-volume applications.

Sample, store and ship small-volume, valuable reagents using Thermo Scientific™ Nalgene Clear PETG Diagnostic Bottles. These bottles are ideal for use in test kits and in many sterile, small-volume applications.

Features:

  • Polyethylene Terephthalate (PETG) with excellent clarity for viewing contents
  • Non-pyrogenic and non-cytotoxic
  • Includes lined, white, high-density polyethylene (HDPE) closure for a leakproof† seal

Ordering Information:

Serum vials are packed in shrink-wrapped trays with closures attached.

Specifications

Capacity 0.16 oz., 5 mL
Capacity (Metric) 5 mL
Capacity (English) 0.16 oz.
Color Clear
Diameter (Metric) Inner Neck 11 mm
Material PETG
No. per Case 500
No. per Pack 100
Sterility Sterile
Neck Finish 20-415 mm
Certifications/Compliance Manufactured in compliance with ISO 13485:2016 quality requirements. Serum Vials meet the requirements of 21CFR177.1315 for food and beverage use, USP Class VI.
Closure Included Yes
Graduated No
Mouth Narrow
Shape Round
Height (Metric) 44 mm
Product Type Narrow Mouth Bottle
Width (English) 0.876 in.
Height (English) 0.45 in.
Closure Liner With Liner
Closure Material HDPE
Closure Size 20-415 mm
Diameter (Metric) 22 mm
Closure Type Cap
Through the Closure Dispensing No
Show More Show Less

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.