Learn More
Thermo Scientific™ Nalgene™ Clear PETG Diagnostic Bottles with Closure: Sterile, Bulk Pack

Description
Sample, store and ship small-volume, valuable reagents using Thermo Scientific™ Nalgene Clear PETG Diagnostic Bottles. These bottles are ideal for use in test kits and in many sterile, small-volume applications.
- Polyethylene Terephthalate (PETG) with excellent clarity for viewing contents
- Non-pyrogenic and non-cytotoxic
- Includes lined, white, high-density polyethylene (HDPE) closure for a leakproof† seal
Ordering Information:
Serum vials are packed in shrink-wrapped trays with closures attached.
Specifications
Specifications
| Capacity | 0.16 oz., 5 mL |
| Capacity (Metric) | 5 mL |
| Capacity (English) | 0.16 oz. |
| Color | Clear |
| Diameter (Metric) Inner Neck | 11 mm |
| Material | PETG |
| No. per Case | 500 |
| No. per Pack | 100 |
| Sterility | Sterile |
| Neck Finish | 20-415 mm |
| Show More |
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.