missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Certified Low Particulate Narrow-Mouth HDPE Bottles with Closure: Lab Pack
GREENER_CHOICE

Catalog No. 0331387B
Change view
Click to view available options
Capacity (Metric):
250 mL
500 mL
1 L
Closure Size:
24 mm
28 mm
38-430 mm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Closure Size
0331387B 250 mL 24 mm
0331387C 500 mL 28 mm
0331387D 1 L 38-430 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 0331387B Supplier Thermo Scientific™ Supplier No. 3820990250
Only null left

Effectively store high-purity chemicals with these HDPE bottles, which are extremely low in trace metal content.

Thermo Scientific™ Nalgene Certified Low Particulate Narrow-Mouth HDPE Bottles are extremely low in trace metal content, making them an excellent choice for storing high-purity chemicals. Controlled environment manufacturing minimizes particulate contamination. Narrow-mouth bottles with certified low-particulate levels are used by shippers who design and assemble combination packaging for high-purity chemicals.

Features:

  • Leakproof†

Includes:

Polypropylene closures

Recommended for:

Storage and shipping of microelectronic or biological reagents

A sample product certificate providing Cytotoxicity, USP Class VI, FDA Compliance—21 CFR and other regulatory compliance information is available or can be requested from our 

Specifications

Capacity 8 oz., 250 mL
Capacity (Metric) 250 mL
Capacity (English) 8 oz.
Color Natural
Description Low-Particulate Narrow-Mouth Bottles
Diameter (Metric) Inner Neck 18 mm
Material HDPE
No. per Case 72
Sterility Non-sterile
Closure Color Natural
Low Particulate Yes
Neck Finish 24 mm
Certifications/Compliance Manufactured in compliance with ISO 13485:2016 quality requirements. Bottles and closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI. Bottles are evaluated at 15psi (103kPa) per 49 CFR 173.27(c)(2), ICAO Technical Instructions Part 4; 1.1.6, and IATA Dangerous Goods Regulations Section 5.0.2.9.
Closure Included Yes
Mouth Narrow
Shape Round
Height (Metric) 133 mm
Product Type Narrow Mouth Bottle
Height (English) 5.23 in.
Closure Material Polypropylene
Closure Size 24 mm
Diameter (Metric) 61 mm
Closure Type Screw Closure (Assembled)
Through the Closure Dispensing No
Show More Show Less

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.