missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ PC Platinum Certified Clean™ Biotainer™ Bottle
GREENER_CHOICE

Catalog No. 14388043
Change view
Click to view available options
Capacity (Metric):
5 mL
20 mL
10 L
Shape:
Round
Square
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Shape
14388043 5 mL Round
14388044 20 mL Round
14388040 10 L Square
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 14388043 Supplier Thermo Scientific™ Supplier No. CEN350005

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Thermo Scientific™ Nalgene™ PC Platinum Certified Clean™ Biotainer™ Bottles are certified and lot-to-lot tested to contain less than 1/3 the particulates allowable according to USP <788> to limit potential particulate contamination.

Products are washed with USP Water for Injection and handled post-washing in Class 5 clean rooms with Class 4 packaging/inspection hoods to produce these very low particulate products. Certified and lot-to-lot tested to contain less than 1/3 the particulates allowable according to USP <788> to limit potential particulate contamination. Sterile, ready-to-use products with a Sterility Assurance Level of 10-6 inside the container/closure, including two heat-sealed sterile barriers to protect product prior to use.

  • Reduce the risk of carry-over contamination and eliminate the cost of cleaning, sterilizing and associated validations
  • Durable, translucent PC construction ensures the integrity of your sample
  • Designed specifically for pharmaceutical and biological manufacturers
  • Closures feature a silicone liner for leakproof assurance
  • Extensive validation documentation support
  • Excellent for freezing applications

Specifications

Capacity (Metric) 5 mL
Capacity (English) 0.16 oz.
Color Transparent Blue
Diameter (Metric) Inner Neck 20 mm
For Use With (Application) Media/Buffer Preparation ​, Media/Buffer Storage, Sampling, Storage, Transportation ​, Validation Testing, Drug Substance, Drug Product
Material Polycarbonate
No. per Case 416/Cs.
No. per Pack 52/Pk.
Sterility Sterile
Temperature Range (Metric) -100°C to 100°C
Weight (English) 9 lb.
Closure Color White
Low Particulate <1/3 USP <788> (Ultralow particulate)
Certifications/Compliance USP Class VI, USP 661, FDA 21 CFR 177.1315
Closure Included Yes
Mouth Narrow
Shape Round
Height (Metric) 46 mm
Product Type Media Bottle
Width (English) 12.5 in.
Closure Liner Silicone
Closure Material Polypropylene
Closure Size 20 mm
Diameter (Metric) 22 mm
Autoclavable Not Autoclavable
Show More Show Less