missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Biohazardous Waste Containers
GREENER_CHOICE

Catalog No. 1181520
Change view
Click to view available options
Capacity (Metric):
5.5 L
19 L
57 L
Diameter (Metric) Outer:
22 cm
28 cm
34 cm
Height (Metric):
28 cm
43 cm
74 cm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Diameter (Metric) Outer Height (Metric)
1181520 5.5 L 22 cm 28 cm
1429363A 19 L 28 cm 43 cm
1429363B 57 L 34 cm 74 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 1181520 Supplier Thermo Scientific™ Supplier No. 63700004

Conduct secondary containment of biohazardous waste with these biohazardous waste containers, which feature convenient one-step sterilization and enable one-handed disposal of materials.

Conduct secondary containment of biohazardous waste with Thermo Scientific™ Nalgene™ Biohazardous Waste Containers, which feature convenient one-step sterilization—no need to remove the bag. A port opening in the cover allows for one-handed disposal of materials, limiting exposure to infectious waste. The containers hold standard sized autoclave bags (not included).

Features:

  • Will not rust, dent or puncture like other materials
  • Complies with U.S. OSHA Standard 29 CFR Part 1910.1030 for use as protection against bloodborne pathogens
  • Autoclavable/Biohazard
  • Symbol printed in English and Spanish

.

Specifications

Capacity (English) 1.5 gal.
Certifications/Compliance U.S. OSHA Standard 29 CFR Part 1910.1030
Diameter (English) Outer 8.75 in.
Capacity (Metric) 5.5 L
Material Polypropylene
Sterility Non-sterile
Type Waste Container Biohazardous
Height (English) 11 in.
Height (Metric) 28 cm
No. per Case 1
No. per Pack 1
Diameter (Metric) Outer 22 cm
Show More Show Less