missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Naked1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113280
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113280 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113280 Supplier Invitrogen™ Supplier No. PA5113280
Only null left

Rabbit Polyclonal Antibody

Naked1 Polyclonal Antibody for Western Blot, ICC/IF, ELISA

In the mouse, Nkd is a Dishevelled (see DVL1; MIM 601365)-binding protein that functions as a negative regulator of the Wnt (see WNT1; MIM 164820)-beta-catenin-Tcf signaling pathway.

Specifications

Antigen Naked1
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 3.52 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene NKD1
Gene Accession No. Q969G9
Gene Alias 2810434J10Rik; 9030215G15Rik; Dvl-binding protein; hNkd; hNkd1; mNkd; mNkd1; naked cuticle 1 homolog; naked cuticle 1 homolog (Drosophila); naked cuticle homolog 1; naked cuticle homolog 1 (Drosophila); naked cuticle-1; Naked1; naked-1; Nkd; NKD1; PP7246; Protein naked cuticle homolog 1
Gene Symbols NKD1
Host Species Rabbit
Immunogen Recombinant Human Protein naked cuticle homolog 1 protein (269-384AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 364952, 85407
Target Species Human, Rat
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less