missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NAGK Monoclonal Antibody (K1E005_8C10), Invitrogen™

Catalog No. MA555856
Change view
Click to view available options
Quantity:
50 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA555856 50 μL
MA556236 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA555856 Supplier Invitrogen™ Supplier No. MA555856
Only null left

Mouse Monoclonal Antibody

Sequence of this protein is as follows: MAAIYGGVEG GGTRSEVLLV SEDGKILAEA DGLSTNHWLI GTDKCVERIN EMVNRAKRKA GVDPLVPLRS LGLSLSGGDQ EDAGRILIEE LRDRFPYLSE SYLITTDAAG SIATATPDGG VVLISGTGSN CRLINPDGSE SGCGGWGHMM GDEGSAYWIA HQAVKIVFDS IDNLEAAPHD IGYVKQAMFH YFQVPDRLGI LTHLYRDFDK CRFAGFCRKI AEGAQQGDPL SRYIFRKAGE MLGRHIVAVL PEIDPVLFQG KIGLPILCVG SVWKSWELLK EGFLLALTQG REIQAQNFFS SFTLMKLRHS SALGGASLGA RHIGHLLPMD YSANAIAFYS YTFS

SPG20 is a protein containing a MIT (Microtubule Interacting and Trafficking molecule) domain, and is implicated in regulating endosomal trafficking and mitochondria function. The protein localizes to mitochondria and partially co-localizes with microtubules. Stimulation with epidermal growth factor (EGF) results in protein translocation to the plasma membrane, and the protein functions in the degradation and intracellular trafficking of EGF receptor.

Specifications

Antigen NAGK
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone K1E005_8C10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Gene NAGK
Gene Accession No. Q9UJ70
Gene Alias GlcNAc kinase; Gnk; HSA242910; N-acetyl-D-glucosamine kinase; N-acetylglucosamine kinase; Nagk
Gene Symbols NAGK
Host Species Mouse
Immunogen Recombinant human NAGK (immunogen range is 1-344).
Purification Method Protein A
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55577
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less