Learn More
Thermo Scientific™ Nafcillin sodium salt
Chemical Identifiers
| CAS | 985-16-0 |
|---|---|
| Molecular Formula | C21H21N2NaO5S |
| Molecular Weight (g/mol) | 436.46 |
| MDL Number | MFCD00056863 |
| InChI Key | AYAPZOUDXCDGIF-FRFVDRIFSA-M |
| Synonym | 4-thia-1-azabicyclo 3.2.0 heptane-2-carboxylicacid, 6-2-ethoxy-1-naphthalenyl carbonyl amino-3,3-dimethyl-7-oxo-, sodiumsalt 1:1 , 2s,5r,6r |
| PubChem CID | 131675018 |
| IUPAC Name | (2S,5R,6R)-6-[(2-ethoxynaphthalene-1-carbonyl)amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid;molecular hydrogen;sodium |
| SMILES | [Na+].CCOC1=CC=C2C=CC=CC2=C1C(=O)N[C@H]1[C@H]2SC(C)(C)[C@@H](N2C1=O)C([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC461380010
|
Thermo Scientific™
461380010 |
1g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC461380050
|
Thermo Scientific™
461380050 |
5g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 985-16-0 | |
| 436.46 | |
| AYAPZOUDXCDGIF-FRFVDRIFSA-M | |
| 131675018 | |
| [Na+].CCOC1=CC=C2C=CC=CC2=C1C(=O)N[C@H]1[C@H]2SC(C)(C)[C@@H](N2C1=O)C([O-])=O |
| C21H21N2NaO5S | |
| MFCD00056863 | |
| 4-thia-1-azabicyclo 3.2.0 heptane-2-carboxylicacid, 6-2-ethoxy-1-naphthalenyl carbonyl amino-3,3-dimethyl-7-oxo-, sodiumsalt 1:1 , 2s,5r,6r | |
| (2S,5R,6R)-6-[(2-ethoxynaphthalene-1-carbonyl)amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid;molecular hydrogen;sodium |
Specifications
| 985-16-0 | |
| C21H21N2NaO5S | |
| 1g | |
| Solubility in water: .. Other solubilities: soluble in methanol | |
| [Na+].CCOC1=CC=C2C=CC=CC2=C1C(=O)N[C@H]1[C@H]2SC(C)(C)[C@@H](N2C1=O)C([O-])=O | |
| 436.46 | |
| 436.46 |
| Authentic | |
| MFCD00056863 | |
| 4-thia-1-azabicyclo 3.2.0 heptane-2-carboxylicacid, 6-2-ethoxy-1-naphthalenyl carbonyl amino-3,3-dimethyl-7-oxo-, sodiumsalt 1:1 , 2s,5r,6r | |
| AYAPZOUDXCDGIF-FRFVDRIFSA-M | |
| (2S,5R,6R)-6-[(2-ethoxynaphthalene-1-carbonyl)amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid;molecular hydrogen;sodium | |
| 131675018 | |
| Nafcillin sodium salt |
Safety and Handling
GHS H Statement May cause an allergic skin reaction. May cause allergy or asthma symptoms or breathing difficulties if inhaled.
GHS P Statement Wear protective gloves/protective clothing. IF ON SKIN: Wash with plenty of soap and water. If skin irritation or rash occurs: Get medical advice/attention. In case of inadequate ventilation wear respiratory protection. IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing. If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
GHS Signal Word: Danger
EINECSNumber : 213-574-4
RUO â Research Use Only