missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nadifloxacin 98.0+%, TCI America™
$121.28 - $358.08
Chemical Identifiers
| CAS | 124858-35-1 |
|---|---|
| Molecular Formula | C19H21FN2O4 |
| Molecular Weight (g/mol) | 360.385 |
| MDL Number | MFCD00865081 |
| InChI Key | JYJTVFIEFKZWCJ-UHFFFAOYSA-N |
| Synonym | 9-Fluoro-8-(4-hydroxy-1-piperidinyl)-5-methyl-1-oxo-1,5,6,7-tetrahydropyrido[3,2,1-ij]quinoline-2-carboxylic Acid |
| PubChem CID | 4410 |
| ChEBI | CHEBI:31889 |
| SMILES | CC1CCC2=C3N1C=C(C(=O)C3=CC(=C2N4CCC(CC4)O)F)C(=O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
N0931200MG
|
TCI America
N0931200MG |
200 mg |
Each for $121.28
|
|
|||||
|
N09311G
|
TCI America
N09311G |
1 g |
Each for $358.08
|
|
|||||
Chemical Identifiers
| 124858-35-1 | |
| 360.385 | |
| JYJTVFIEFKZWCJ-UHFFFAOYSA-N | |
| 4410 | |
| CC1CCC2=C3N1C=C(C(=O)C3=CC(=C2N4CCC(CC4)O)F)C(=O)O |
| C19H21FN2O4 | |
| MFCD00865081 | |
| 9-Fluoro-8-(4-hydroxy-1-piperidinyl)-5-methyl-1-oxo-1,5,6,7-tetrahydropyrido[3,2,1-ij]quinoline-2-carboxylic Acid | |
| CHEBI:31889 |
Specifications
| 124858-35-1 | |
| White-Yellow | |
| MFCD00865081 | |
| 9-Fluoro-8-(4-hydroxy-1-piperidinyl)-5-methyl-1-oxo-1,5,6,7-tetrahydropyrido[3,2,1-ij]quinoline-2-carboxylic Acid | |
| CC1CCC2=C3N1C=C(C(=O)C3=CC(=C2N4CCC(CC4)O)F)C(=O)O | |
| 4410 | |
| 360.39 | |
| Crystalline Powder |
| 247°C | |
| C19H21FN2O4 | |
| 200 mg | |
| JYJTVFIEFKZWCJ-UHFFFAOYSA-N | |
| 360.385 | |
| CHEBI:31889 | |
| ≥98.0% (HPLC,T) | |
| Nadifloxacin |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator