missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Nadifloxacin | |
| 247°C | |
| C19H21FN2O4 | |
| 1 g | |
| JYJTVFIEFKZWCJ-UHFFFAOYSA-N | |
| 360.385 | |
| CHEBI:31889 | |
| ≥98.0% (HPLC,T) |
| 124858-35-1 | |
| White-Yellow | |
| MFCD00865081 | |
| 9-Fluoro-8-(4-hydroxy-1-piperidinyl)-5-methyl-1-oxo-1,5,6,7-tetrahydropyrido[3,2,1-ij]quinoline-2-carboxylic Acid | |
| CC1CCC2=C3N1C=C(C(=O)C3=CC(=C2N4CCC(CC4)O)F)C(=O)O | |
| 4410 | |
| 360.39 | |
| Crystalline Powder |
Chemical Identifiers
| 124858-35-1 | |
| 360.385 | |
| JYJTVFIEFKZWCJ-UHFFFAOYSA-N | |
| 4410 | |
| CC1CCC2=C3N1C=C(C(=O)C3=CC(=C2N4CCC(CC4)O)F)C(=O)O |
| C19H21FN2O4 | |
| MFCD00865081 | |
| 9-Fluoro-8-(4-hydroxy-1-piperidinyl)-5-methyl-1-oxo-1,5,6,7-tetrahydropyrido[3,2,1-ij]quinoline-2-carboxylic Acid | |
| CHEBI:31889 |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator